Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adiponectin/Acrp30 Antibody (OTI1H1), Alexa Fluor™ 350, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP271422AF350

Catalog No. NP271422A30


Only null left
Add to Cart

Description

Description

Adiponectin/Acrp30 Monoclonal specifically detects Adiponectin/Acrp30 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Adiponectin/Acrp30
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
ADIPOQ
Recombinant protein fragment corresponding to amino acids 19-244 of human Adiponectin/Acrp30 (NP_004788) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1H1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein
Mouse
Affinity Purified
Diabetes Research, Lipid and Metabolism
9370.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.