Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adiponectin/Acrp30 Rat anti-Mouse, Alexa Fluor 532, Clone: MADI 04, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280031AF532
This item is not returnable.
View return policy
Description
Adiponectin/Acrp30 Monoclonal antibody specifically detects Adiponectin/Acrp30 in Mouse samples. It is validated for Western Blot, ELISASpecifications
Adiponectin/Acrp30 | |
Monoclonal | |
Alexa Fluor 532 | |
ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein | |
Recombinant mouse Adiponectin/Acrp30. | |
0.1 mL | |
Diabetes Research, Lipid and Metabolism | |
9370 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
MADI 04 | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction