Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adiponectin Mouse anti-Human, Clone: OTI3F12, lyophilized, TrueMAB™
SDP

Catalog No. 501667032
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-7032 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-7032 Supplier OriGene Supplier No. CF804554
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Adiponectin is a hormone exclusively produced in adipose tissue and secreted into plasma. It is involved in a number of metabolic processes, such as glucose regulation, and the metabolism of fat for energy production. Adiponectin is involved in the control of fat metabolism and insulin sensitivity.

Specifications

Antigen Adiponectin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3F12
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene ADIPOQ
Gene Accession No. Q15848
Gene Alias ACDC, ACRP30, ADIPQTL1, ADPN, APM-1, APM1, GBP28
Gene Symbols ADIPOQ
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 19-244 of human ADIPOQ produced in SF9 cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9370
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.