Learn More
Invitrogen™ Adiponectin Polyclonal Antibody

Description
Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. In Western blot, this antibody detects a band at ~26.4kDa in human HL60 cell lysates. A suggested positive control for PA1-84881 is rat brain. Rabbit anti Human adiponectin antibody recognizes an epitope within the C-terminal region (CT) of the 244 amino acid protein adiponectin (also known as Acrp30), a major adipokine secreted into the bloodstream from adipose tissue to modulate metabolism, including glucose regulation and fatty acid catabolism.
Specifications
Specifications
| Antigen | Adiponectin |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ADIPOQ |
| Gene Accession No. | Q15848, Q60994 |
| Gene Alias | 30 kDa adipocyte complement-related protein; 30kDa; Acdc; Acrp30; Ad; adipo; adipocyte complement related protein; adipocyte complement related protein of 30 kDa; adipocyte complement-related 30 kDa protein; adipocyte complement-related protein of 30 kDa; adipocyte, C1Q and collagen domain containing; adipocyte, C1q and collagen domain-containing protein; Adipocyte-specific protein AdipoQ; adiponectin; adiponectin, C1Q and collagen domain containing; Adipoq; Adipose most abundant gene transcript 1 protein; adipose specific collagen-like factor; ADIPQTL1; ADPIOQ; ADPN; Apm1; APM-1; APN; GBP28; Gelatin-binding protein; gelatin-binding protein 28 |
| Gene Symbols | ADIPOQ |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.