Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Adiponectin, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABS516AF488 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS516AF488 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS516AF488 Supplier MilliporeSigma™ Supplier No. ABS516AF488

Rabbit Polyclonal Antibody

Adiponectin (UniProt Q8K3R4; also known as 30kDa adipocyte complement-related protein, ACRP30, Adipocyte complement-related 30kDa protein, Adipocyte, C1q and collagen domain-containing protein, Adipose most abundant gene transcript 1 protein, apM-1, Gelatin-binding protein) is encoded by the Adipoq (also known as Acdc, Acrp30) gene (Gene ID 246253) in rat. Adipokines are adipose tissue-secreted factors involved in vascular function and metabolic disease. While most adipokines are proinflammatory and directly responsible for the development of the chronic inflammatory state in obesity, adiponectin is an adipokine with anti-inflammatory and insulin-sensitizing properties. Adiponectin mediates it biological activity via adiponection receptor protein 1 and 2 (AdipoR1 and AdipoR2). Both AdipoR1 and AdipoR2 are reported to be ubiquitously expressed with the highest expression in skeletal muscle and liver, respectively. Circulating adiponectin levels are reduced in obesity and type II diabetes, whereas weight loss elevates serum adiponectin. Likewise, adiponectin overexpression is reported to alleviate metabolic dysfunction independent of weight loss in both genetic and diet-induced animal models of obesity. Aadiponectin deficiency is associated with various clinical cardiovascular disorders including myocardial infarction, peripheral artery disease, and endothelial dysfunction. Adiponectin is produced with a signal peptide sequence (a.a. 1-18 in human and rat sequence; a.a. 1-17 in mouse), which is removed postranslationally to yield the mature protein with a collagen-like domain (a.a. 42-107 in human and rat, a.a. 45-110 in mouse) and a C1q domain (a.a. 108-244 in human and rat, a.a. 111-247 in mouse).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adiponectin
Applications Immunofluorescence
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody conjugate in PBS with 1.5% BSA and 0.05% Sodium Azide.
Gene Adipoq, Acdc, Acrp30
Gene Accession No. Q8K3R4
Host Species Rabbit
Immunogen GST-tagged recombinant rat adiponectin collagen-like domain.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_653345
Test Specificity This Alexa Fluor™ 488 conjugate was prepared from purified rabbit polyclonal antibody (Mfr. No. ABS516) that was raised against rat adiponectin collagen-like domain and validated for target band detection in both rat and mouse serum samples by Western blotting. Human and Rat adiponectins share 82% sequence identity in the collagen-like domain.
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.