Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adiponectin receptor 1 Mouse anti-Human, Clone: 1F12B3, Proteintech
SDP

Catalog No. 501737037 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7037 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7037 Supplier Proteintech Group Inc Supplier No. 666191IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody was raised against 1-72 aa of human AdipoR1.

The adiponectin receptors, ADIPOR1 and ADIPOR2, serve as receptors for globular and full-length adiponectin and mediate increased AMPK and PPAR-alpha ligand activities, as well as fatty acid oxidation and glucose uptake by adiponectin.

Specifications

Antigen Adiponectin receptor 1
Applications Western Blot
Classification Monoclonal
Clone 1F12B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ADIPOR1
Gene Accession No. Q96A54
Gene Alias ACDCR1, Adiponectin receptor 1, Adiponectin receptor protein 1, ADIPOR1, CGI 45, CGI45, PAQR1, TESBP1A
Gene Symbols ADIPOR1
Host Species Mouse
Immunogen Adiponectin receptor 1 Fusion Protein Ag18187
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51094
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.