Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adiponectin receptor 2 Polyclonal Antibody, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS0611R

This item is not returnable.
View return policy
Description
Adiponectin receptor 2 Polyclonal antibody specifically detects Adiponectin receptor 2 in Human, Mouse, Rat samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
Adiponectin receptor 2 | |
Polyclonal | |
Unconjugated | |
ADIPOR2 | |
1110001I14Rik; ACDCR2; ADCR2; adiponectin receptor 2; adiponectin receptor protein 2; Adipor2; AI115388; AW554121; D6Ucla1e; FLJ21432; MGC4640; Paqr2; Parq2; Progestin and adipoQ receptor family member 2; Progestin and adipoQ receptor family member II | |
KLH conjugated synthetic peptide derived from human Adiponectin receptor 2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
Q86V24, Q8BQS5 | |
Rabbit | |
Protein A | |
RUO | |
312670, 68465, 79602 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction