Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adiponitrile, 98%

Catalog No. AAB23122AK
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB23122AK 250 mL
AAB23122AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB23122AK Supplier Thermo Scientific Chemicals Supplier No. B23122AK
Only null left
Add to Cart

CAS: 111-69-3 | C6H8N2 | 108.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-69-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00001975
InChI Key BTGRAWJCKBQKAO-UHFFFAOYSA-N
Synonym adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril
PubChem CID 8128
IUPAC Name hexanedinitrile
SMILES C(CCC#N)CC#N

Specifications

Melting Point 2°C
Density 0.951
Boiling Point 295°C
Flash Point 163°C (325°F)
Odor Odorless
Refractive Index 1.438
Quantity 250 mL
UN Number UN2205
Beilstein 1740005
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material Adiponitrile
Show More Show Less
Hazard Category H301-H332
Hazard Statement GHS H Statement
H301-H331-H227
Toxic if swallowed.
Toxic if inhaled.
Combustible liquid.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ADIPONITRILE
EINECSNumber 203-896-3
RTECSNumber AV2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.