Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Adiponitrile, 98%

Click to view available options
Quantity:
250mL
1000mL
111-69-3
C6H8N2
108.144
MFCD00001975
BTGRAWJCKBQKAO-UHFFFAOYSA-N
adiponitrile,1,4-dicyanobutane,adipodinitrile,adipyldinitrile,adipic acid dinitrile,tetramethylene dicyanide,nitrile adipico,tetramethylene cyanide,adipic acid nitrile,adipinsaeuredinitril
8128
hexanedinitrile
C(CCC#N)CC#N
111-69-3
C6H8N2
108.144
MFCD00001975
BTGRAWJCKBQKAO-UHFFFAOYSA-N
adiponitrile,1,4-dicyanobutane,adipodinitrile,adipyldinitrile,adipic acid dinitrile,tetramethylene dicyanide,nitrile adipico,tetramethylene cyanide,adipic acid nitrile,adipinsaeuredinitril
8128
hexanedinitrile
C(CCC#N)CC#N
Boiling Point 295°C
Melting Point 2°C
Quantity 1000mL
Odor Odorless
Flash Point 163°C (325°F)
UN Number UN2205
Beilstein 1740005
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material Adiponitrile

GHS H Statement
H301-H331-H227
Toxic if swallowed.
Toxic if inhaled.
Combustible liquid.

GHS P Statement
P210-P261-P280-P301+P310-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ADIPONITRILE

RTECSNumber : AV2625000

TSCA : TSCA