Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adiponutrin/PNPLA3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP100227
Description
Adiponutrin/PNPLA3 Polyclonal specifically detects Adiponutrin/PNPLA3 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Adiponutrin/PNPLA3 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Acylglycerol O-acyltransferase, adiponutrin, ADPNdJ796I17.1, C22orf20, Calcium-independent phospholipase A2-epsilon, chromosome 22 open reading frame 20, EC 2.3.1.-, EC 2.7.7.56, EC 3.1.1.3, EC 4.2.3.4, FLJ22012, iPLA(2)epsilon, iPLA2-epsilon, patatin-like phospholipase domain containing 3, patatin-like phospholipase domain-containing protein 3 | |
Goat | |
Affinity Purified | |
RUO | |
80339 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.01 - 0.03 μg/mL, Immunohistochemistry-Paraffin 8 μg/mL, Peptide ELISA 1:16000 | |
NP_473429.2 | |
PNPLA3 | |
Peptide with sequence EHDICPKVKSTN corresponding to internal region according to Uniprot Mouse NP_473429.2. | |
0.1 mg | |
Primary | |
This antibody is raised against a peptide derived from the Mouse protein sequence and this peptide differs from Human only in position 2. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction