Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AdipoR1 Antibody (2416C), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10128MFV610
Description
AdipoR1 Monoclonal antibody specifically detects AdipoR1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
AdipoR1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
0.1 mL | |
Signal Transduction | |
51094 | |
Store at 4C in the dark. |
Flow Cytometry, Immunohistochemistry, CyTOF | |
2416C | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
ACDCR1, adiponectin receptor 1, adiponectin receptor protein 1, ADR1, CGI45, FLJ42464, PAQR1FLJ25385, Progestin and adipoQ receptor family member I, TESBP1A | |
Synthetic peptide containing human AdipoR1 C-term sequences, | |
RUO | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction