Learn More
Description
- Agonist of adiponectin receptors AdipoR1 and AdipoR2 (Kd values are 1.8 and 3.1 μm respectively).
- Activates AMPK and PPARα signaling; ameliorates insulin resistance, dyslipidemia and glucose intolerance in db/db mice.
- Orally active antidiabetic agent.
Specifications
Specifications
| Chemical Name or Material | 2-(4-Benzyoylphenoxy)-N-[1-(phenylmethyl)-4-piperidinyl]acetamide hydrochloride |
| CAS | 1781835-20-8 |
| Quantity | 10 mg |
| Molecular Formula | C27H28N2O3.HCl |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

