Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Adjustable Bouffant Cap
SDP

Catalog No. p-7240064
Click to view available options
Color:
Green
This item is not returnable. View return policy
This item is not returnable. View return policy

Bouffant caps keeps hair contained and are ideal for food processing, manufacturing, or general industry.

  • Polypropylene
  • 100% Latex Free
  • Economical protection
  • Manufactured in the USA

Specifications

Product Type Bouffant
Material Polypropylene
Disposable Single-use
Latex Free Yes
Model 110NW
Sterility Non-sterile
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Style Traditional
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.