Learn More
Thermo Scientific™ Adjustable Test Tube Racks with Holder for MaxQ™ High Performance Shakers
Catalog No. p-4531054
Click to view available options
Holds:
10-13 mm Tubes
16-20mm Tubes
21-25 mm Tubes
26-30 mm Tubes
Place these adjustable rack holders at any angle to increase sample aeration. They offer exceptional flexibility and incredible convenience for 10 to 30 mm diameter tubes.
Place these adjustable rack holders at any angle to increase sample aeration. Thermo Scientific™ Adjustable Test Tube Racks with Holder for MaxQ™ 8000 Shakers provide added convenience and will accommodate test tube diameters from 10 to 30 mm. These rack holders deliver exceptional flexibility — the right choice for shaking tubes.
  • Can be used only with their corresponding rack size
Includes: One test tube rack
TRUSTED_SUSTAINABILITY
Specifications
Holds 10-13 mm Tubes
Tube Size 10 to 13 mm
Tube Capacity 3 mL to 7 mL
Product Type Test Tube Rack with Holder
Description Adjustable test tube rack
No. of Positions 72
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.