Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADK Antibody (4F8), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197934AF405
Description
ADK Monoclonal specifically detects ADK in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ADK | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ADK | |
Recombinant human ADK (22-362aa) purified from E. coli | |
100 μL | |
Lipid and Metabolism | |
132 | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
4F8 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
adenosine kinase, AKAdenosine 5'-phosphotransferase, EC 2.7.1, EC 2.7.1.20 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human ADK mAb, clone 4F8, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ADK protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction