Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578731
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578731 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578731 Supplier Invitrogen™ Supplier No. PA578731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Human Stefin B shares 78.6 % amino acid (aa) sequence identity with both mouse and rat Stefin B. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human PC-3 whole cell, human T47D whole cell, human U937 whole cell, human U251 whole cell, human HepG2 whole cell, rat liver tissue, rat kidney tissue, rat stomach tissue, rat PC-12 whole cell, mouse kidney tissue, mouse stomach tissue, mouse NIH/3T3 whole cell. IHC: human placenta tissue, mouse liver tissue, rat liver tissue. ICC/IF: PC-3 cell.

The protein encoded by this gene catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene ADK
Gene Accession No. P55263, P55264, Q64640
Gene Alias 2310026J05Rik; 5033405D03Rik; Adenosine 5'-phosphotransferase; adenosine kinase; ADK; AI255373; AI987814; AK; testicular tissue protein Li 14
Gene Symbols ADK
Host Species Rabbit
Immunogen E. coli-derived human ADK recombinant protein (Position: K165-T351).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11534, 132, 25368
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.