Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADM receptor Polyclonal Antibody, Biotin, Bioss
SDP

Catalog No. 502280197
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0197 Supplier Bioss Supplier No. BS22054RBIOTIN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ADM receptor Polyclonal antibody specifically detects ADM receptor in Mouse samples. It is validated for Western Blot

Specifications

Antigen ADM receptor
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene GPR182
Gene Accession No. P43142
Gene Alias 7TMR; ADMR; adrenomedullin receptor; AMR; AM-R; G protein-coupled receptor 182; G10D; G10-D; gamrh; Gpcr17; Gpcr22; GPR182; G-protein coupled receptor 182; HAMR; hrhAMR; MB10; NOW; RAMR
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse ADM receptor
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11536
Target Species Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.