Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550482
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550482 Supplier Invitrogen™ Supplier No. PA550482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ADM2 protein.

This gene encodes a protein which is a member of the calcitonin-related hormones. The encoded protein is involved in maintaining homeostasis in many tissues, acting via CRLR/RAMP receptor (calcitonin receptor-like receptor/receptor activity-modifying protein) complexes. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADM2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ADM2
Gene Accession No. Q7Z4H4
Gene Alias ADM2; adrenomedullin 2; Adrenomedullin-2; AM2; dJ579N16.4; Imd; IMDL; Imdn; IMDS; Intermedin; Intermedin-long; Intermedin-short; Protein ADM2
Gene Symbols ADM2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human adrenomedullin 2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79924
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.