Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADNP Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297747JF646
Description
ADNP Polyclonal specifically detects ADNP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ADNP | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
Activity-dependent neuroprotective protein, activity-dependent neuroprotector, activity-dependent neuroprotector homeobox, ADNP homeobox 1, ADNP1KIAA0784activity-dependent neuroprotector homeobox protein | |
Produced in rabbits immunized with E. coli-derived Human ADNP fragment. | |
0.1 mL | |
Cancer, Neuroscience | |
23394 | |
Store at 4°C in the dark. | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction