Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADNP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14223
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14223 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14223 Supplier Invitrogen™ Supplier No. PA14223
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4223 detects the hADNP protein in human samples. PA1-4223 has been successfully used in Western blot procedures. The PA1-4223 immunogen is a synthetic peptides rcorresponding to residues M(1) F Q L P V N(7) C K(1096) L S S Q Q A(1102) of the N- and C-termini of human ADNP, separated by a cysteine.

Vasoactive intestinal peptide is a neuroprotective factor that has a stimulatory effect on the growth of some tumor cells and an inhibitory effect on others. This gene encodes a protein that is upregulated by vasoactive intestinal peptide and may be involved in its stimulatory effect on certain tumor cells. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor. This gene is also upregulated in normal proliferative tissues. Finally, the encoded protein may increase the viability of certain cell types through modulation of p53 activity. Alternatively spliced transcript variants encoding the same protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADNP
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Adnp
Gene Accession No. Q9H2P0
Gene Alias AA589558; activity dependent neuroprotector homeobox; activity-dependent neuroprotective protein; activity-dependent neuroprotector; activity-dependent neuroprotector homeobox; activity-dependent neuroprotector homeobox protein; Adnp; ADNP homeobox 1; ADNP1; HVDAS; KIAA0784; mKIAA0784; MRD28; NAP peptide
Gene Symbols Adnp
Host Species Rabbit
Immunogen Synthetic peptides rcorresponding to residues M(1) F Q L P V N(7) C K(1096) L S S Q Q A(1102) of the N- and C-termini of human ADNP.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23394
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.