Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADNP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568999
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 93%; Rat: 93%.
ADNP2 gene ontology annotations related to this gene include cellular response to oxidative stress; cellular response to retinoic acid; negative regulation of cell death; neuron differentiation; positive regulation of cell growth.
Specifications
ADNP2 | |
Polyclonal | |
Unconjugated | |
ADNP2 | |
8430420L05Rik; Activity-dependent neuroprotector homeobox protein 2; ADNP homeobox 2; ADNP homeobox protein 2; ADNP homeobox protein 2; activity-dependent neuroprotector homeobox protein 2; ADNP homeobox protein 2-like protein; ADNP2; BC024969; Kiaa0863; mKIAA0863; RGD1308279; Zfp508; Zinc finger protein 508; Znf508 | |
Rabbit | |
Affinity chromatography | |
RUO | |
307236 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
ADNP2 | |
synthetic peptide directed towards the N-terminal region of rat ADNP2. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction