Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADNP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552388 Supplier Invitrogen™ Supplier No. PA552388
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TCPVCNELFP SNVYQVHMEV AHKHSESKSG EKLEPEKLAA CAPFLKWMRE KTVRCLSCKC LVSEEELIHH LLMHGLGCLF CPCTFHDIKG LSEHSRNRHL GKKKLPMDYS NRGFQLDVDA NGNLLFPHLD FITILPKEKL GERE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

ADNP2 gene ontology annotations related to this gene include cellular response to oxidative stress; cellular response to retinoic acid; negative regulation of cell death; neuron differentiation; positive regulation of cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADNP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADNP2
Gene Accession No. Q6IQ32, Q8CHC8
Gene Alias 8430420L05Rik; Activity-dependent neuroprotector homeobox protein 2; ADNP homeobox 2; ADNP homeobox protein 2; ADNP homeobox protein 2; activity-dependent neuroprotector homeobox protein 2; ADNP homeobox protein 2-like protein; ADNP2; BC024969; Kiaa0863; mKIAA0863; RGD1308279; Zfp508; Zinc finger protein 508; Znf508
Gene Symbols ADNP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADNP2. Recombinant protein control fragment (Product #RP-89311).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22850, 240442, 307236
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.