Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADO Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ADO |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:10-1:20 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ADO Polyclonal specifically detects ADO in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ADO | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
84890 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GASDRDAASGPEAPMQPGFPENLSKLKSLLTQLRAEDLNIAPRKATLQPL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:10-1:20 | |
Polyclonal | |
Rabbit | |
Human | |
2-aminoethanethiol (cysteamine) dioxygenase, 2-aminoethanethiol dioxygenase, C10orf22, chromosome 10 open reading frame 22, cysteamine (2-aminoethanethiol) dioxygenase (ADO), Cysteamine dioxygenase, DKFZp564C046, EC 1.13.11.19, FLJ14547 | |
ADO | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title