Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AdoHcyase 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596800
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596800 Supplier Invitrogen™ Supplier No. PA596800
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATVTKAPKK QIQFADDMQE FTKFPTKTGR RSLSRSISQS STDSYSSAAS YTDSSDDEVS PREKQQTNSK GSSNFCVKNI KQAEFGRREI EIAEQDMSAL ISLRKRAQGE KPLAGAKIVG CTHITAQTAV LIETLCALGA QCRWSACNIY STQNEVAAAL AEAGVAVFAW KGESEDDFWW CIDRCVNMDG WQANMILDDG GDLTHWVYKK YPNVFKKIRG IVEESVTGVH; Positive Samples: 293T, U-251MG, SKOV3, Mouse brain, Rat spinal cord; Cellular Location: Apical cell membrane, Cytoplasm, Endoplasmic reticulum, Microsome, cytosol.

The protein encoded by this gene interacts with inositol 1,4,5-trisphosphate receptor, type 1 and may be involved in the conversion of S-adenosyl-L-homocysteine to L-homocysteine and adenosine. Several transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AdoHcyase 2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AHCYL1
Gene Accession No. B5DFN2, O43865, Q80SW1
Gene Alias 1110034F20Rik; AA409031; AA414901; adenosylhomocysteinase 2; adenosylhomocysteinase 2; S-adenosylhomocysteine hydrolase-like protein 1; adenosylhomocysteinase like 1; adenosylhomocysteinase-like 1; adoHcyase 2; Ahcyl1; Ahcyl1-v1; Ahcyl1-v2; Ahcy-rs3; cb586; DCAL; DC-expressed AHCY-like molecule; dendritic cell expressed AHCY-like protein; inositol 1,4,5-trisphosphate receptor-binding protein; IP(3)Rs binding protein released with IP(3); IP3R binding protein released with inositol 1,4,5-trisphosphate; IP3R-binding protein released with inositol 1,4,5-trisphosphate; Irbit; PPP1R78; PRO0233; protein phosphatase 1, regulatory subunit 78; putative adenosylhomocysteinase 2; s adenosylhomocysteine hydrolase; S-adenosyl homocysteine hydrolase homolog; S-adenosylhomocysteine hydrolase, related sequence 3; S-adenosylhomocysteine hydrolase-like 1; S-adenosylhomocysteine hydrolase-like protein 1; S-adenosylhomocysteine hydrolase-like protein variant 1; S-adenosylhomocysteine hydrolase-like protein variant 2; S-adenosyl-L-homocysteine hydrolase 2; wu:fj66f02; XPVKONA
Gene Symbols AHCYL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 31-260 of human AHCYL1 (NP_0066122).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10768, 229709, 362013
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.