Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADORA2A Monoclonal Antibody (6A6G11)
GREENER_CHOICE

Catalog No. PIMA531611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531611 Supplier Invitrogen™ Supplier No. MA531611
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31611 has been tested in indirect ELISA.

ADORA2A is a protein which is one of several receptor subtypes for adenosine. The activity of the encoded protein, a G-protein coupled receptor family member, is mediated by G proteins which activate adenylyl cyclase. This protein is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADORA2A
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6A6G11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene ADORA2A
Gene Accession No. P29274
Gene Alias A2a receptor; A2AAR; A2aR; AA2AR; ADENO; adenosine A2a receptor; Adenosine A2a-receptor; adenosine receptor A2a; adenosine receptor subtype A2a; ADORA2; ADORA2A; Adora2l1; AR; hA2aR; RDC8; unnamed protein product
Gene Symbols ADORA2A
Host Species Mouse
Immunogen Purified recombinant fragment of human ADORA2A (AA: 274-412) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.