Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADORA2A Mouse anti-Human, Clone:6A6G11, Abnova™

Catalog No. 89334040 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-040 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-040 Supplier Abnova Corporation Supplier No. MAB17574
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human ADORA2A.

This gene encodes a protein which is one of several receptor subtypes for adenosine. The activity of the encoded protein, a G-protein coupled receptor family member, is mediated by G proteins which activate adenylyl cyclase. The encoded protein is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine. [provided by RefSeq]

Specifications

Antigen ADORA2A
Applications ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 6A6G11
Conjugate Unconjugated
Description adenosine A2a receptor
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry (1:100-1:500)
Flow Cytometry (1:200-1:400)
Immunohistochemistry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene ADORA2A
Gene Alias ADORA2, RDC8, hA2aR
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 274-412 of human ADORA2A from E. coli.
Quantity 100 μg
Gene ID (Entrez) 135
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.