Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADORA2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577849 Supplier Invitrogen™ Supplier No. PA577849
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS, mouse brain, rat brain Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ADORA2B encodes an adenosine receptor that is a member of the G protein-coupled receptor superfamily. This integral membrane protein stimulates adenylate cyclase activity in the presence of adenosine. This protein also interacts with netrin-1, which is involved in axon elongation. The gene is located near the Smith-Magenis syndrome region on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADORA2B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene ADORA2B
Gene Accession No. P29275, P29276, Q60614
Gene Alias A2b; A2b receptor; A2b, Rs; A2BAR; A2BR; AA2BR; adenosine A2B receptor; adenosine receptor A2b; ADORA2; Adora2b; AI480866; AI605384; HGNC:264; netrin-1 receptor
Gene Symbols ADORA2B
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Extracellular domain of human Adenosine A2b Receptor. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11541, 136, 29316
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.