Learn More
ADORA2B Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 210711AP150UL
Description
The ADORA2B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ADORA2B. This antibody recognizes human, mouse antigen. The ADORA2B antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
ADORA2B | |
Polyclonal | |
Unconjugated | |
150 μL | |
136 | |
Liquid |
Western Blot, ELISA | |
600 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.