Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADORA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570463
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570463 Supplier Invitrogen™ Supplier No. PA570463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Human: 100%; Pig: 79%.

Adenosine is involved in a variety of processes, including the synthesis of urea, the anti-inflammatory response and the inhibition of protein synthesis The adenosine receptors, including adenosine A1-R, adenosine A2A-R, adenosine A2B-R and adenosine A3-R, are integral membrane proteins that are members of the G protein-coupled receptor family. The A1-R protein mediates ureagenesis in a partially calcium-dependent manner. Adenosine is known to mediate coronary vasodilation via the A2A-R receptor. Collagen synthesis and total protein synthesis are inhibited in certain cells by adenosine, acting via the A2B receptors. Activation of the A3-R receptor inhibits the induction of the cytokine TNF alpha and blocks the endotoxin CD14 receptor signal transduction pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADORA3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADORA3
Gene Accession No. P0DMS8
Gene Alias A3 adenosine receptor; A3AR; A3R; AA3R; AD026; adenosine A3 receptor; Adenosine A3-R; adenosine receptor A3; adenosine-3 receptor; Adora3; Agtrl1; angiotensin II receptor-like 1; angiotensin receptor-like 1; Apelin receptor; Apj; APJ (apelin) receptor; APJ receptor; APJR; Aplnr; ARA3; B78; BA552M11.5; G protein-coupled receptor; G protein-coupled receptor APJ; Gpcr2; GPCR34; G-protein coupled receptor 2; G-protein coupled receptor APJ; G-protein coupled receptor HG11; HG11; HG11 orphan receptor; MSR; msr/apj; RP11-552M11.7; TGPCR1
Gene Symbols ADORA3
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human ADORA3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.