Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADORA3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536350
Description
This antibody detects endogenous protein at a molecular weight of 38 kDa. Purity is >95% by SDS-PAGE.
Adenosine is involved in a variety of processes, including the synthesis of urea, the anti-inflammatory response and the inhibition of protein synthesis The adenosine receptors, including adenosine A1-R, adenosine A2A-R, adenosine A2B-R and adenosine A3-R, are integral membrane proteins that are members of the G protein-coupled receptor family. The A1-R protein mediates ureagenesis in a partially calcium-dependent manner. Adenosine is known to mediate coronary vasodilation via the A2A-R receptor. Collagen synthesis and total protein synthesis are inhibited in certain cells by adenosine, acting via the A2B receptors. Activation of the A3-R receptor inhibits the induction of the cytokine TNF alpha and blocks the endotoxin CD14 receptor signal transduction pathway.
Specifications
ADORA3 | |
Polyclonal | |
Unconjugated | |
ADORA3 | |
A3 adenosine receptor; A3AR; A3R; AA3R; AD026; adenosine A3 receptor; Adenosine A3-R; adenosine receptor A3; adenosine-3 receptor; Adora3; Agtrl1; angiotensin II receptor-like 1; angiotensin receptor-like 1; Apelin receptor; Apj; APJ (apelin) receptor; APJ receptor; APJR; Aplnr; ARA3; B78; BA552M11.5; G protein-coupled receptor; G protein-coupled receptor APJ; Gpcr2; GPCR34; G-protein coupled receptor 2; G-protein coupled receptor APJ; G-protein coupled receptor HG11; HG11; HG11 orphan receptor; MSR; msr/apj; RP11-552M11.7; TGPCR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11542, 140, 25370 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P0DMS8, P28647, Q61618 | |
ADORA3 | |
Synthetic peptide corresponding to amino acids C-terminus of human ADORA3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction