Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADORA3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102048
Description
Antibody detects endogenous levels of total ADORA3.
Adenosine is involved in a variety of processes, including the synthesis of urea, the anti-inflammatory response and the inhibition of protein synthesis The adenosine receptors, including adenosine A1-R, adenosine A2A-R, adenosine A2B-R and adenosine A3-R, are integral membrane proteins that are members of the G protein-coupled receptor family. The A1-R protein mediates ureagenesis in a partially calcium-dependent manner. Adenosine is known to mediate coronary vasodilation via the A2A-R receptor. Collagen synthesis and total protein synthesis are inhibited in certain cells by adenosine, acting via the A2B receptors. Activation of the A3-R receptor inhibits the induction of the cytokine TNF alpha and blocks the endotoxin CD14 receptor signal transduction pathway.
Specifications
ADORA3 | |
Polyclonal | |
Unconjugated | |
ADORA3 | |
A3 adenosine receptor; A3AR; A3R; AA3R; AD026; adenosine A3 receptor; Adenosine A3-R; adenosine receptor A3; adenosine-3 receptor; Adora3; Agtrl1; angiotensin II receptor-like 1; angiotensin receptor-like 1; Apelin receptor; Apj; APJ (apelin) receptor; APJ receptor; APJR; Aplnr; ARA3; B78; BA552M11.5; G protein-coupled receptor; G protein-coupled receptor APJ; Gpcr2; GPCR34; G-protein coupled receptor 2; G-protein coupled receptor APJ; G-protein coupled receptor HG11; HG11; HG11 orphan receptor; MSR; msr/apj; RP11-552M11.7; TGPCR1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11542, 140 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P0DMS8, Q61618 | |
ADORA3 | |
A synthesized peptide derived from human ADORA3(Accession P0DMS8), corresponding to amino acid residues Y284-E318. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction