Learn More
Promega ADP-Glo Kinase Assay + AKT2 Kinase Enzyme System, 1 each
Catalog No. PRV9041 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRV9041 1 Ea.
1 options
Catalog No. PRV9041 Supplier Promega Supplier No. V9041
Recombinant full-length human AKT2 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. AKT2 or Protein Kinase B beta (PKB beta) is a serine/threonine kinase that is a member of the AKT family.
  • Easily Screen and Profile AKT2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 Ea.
Content And Storage Components at different temperatures