Learn More
Promega ADP-Glo Kinase Assay + ASK1 Kinase Enzyme System, 1 each
Catalog No. PRV9481 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRV9481 1 Ea.
1 options
Catalog No. PRV9481 Supplier Promega Supplier No. V9481
Recombinant human ASK1 (649-946) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. ASK1, also known as MAPKKK5, activates MKK3, MKK4 (SEK1) and MKK6. Overexpression of ASK1 induces apoptotic cell death.
  • Easily Screen and Profile ASK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 Ea.
Content And Storage Components at different temperatures