Learn More
Promega ADP-Glo Kinase Assay + Aurora A Kinase Enzyme System, 1 each
Catalog No. PRV9081 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRV9081 1 Ea.
1 options
Catalog No. PRV9081 Supplier Promega Supplier No. V9081
Recombinant full-length human Aurora A was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. Aurora A belongs to a multigenic family of mitotic serine/threonine kinases, which are involved in the control of chromosome segregation.
  • Easily Screen and Profile Aurora A Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 Ea.
Content And Storage Components at different temperatures