Learn More
Promega ADP-Glo Kinase Assay + Aurora B Kinase Enzyme System, 1 each
Catalog No. PRV9181 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRV9181 1 Ea.
1 options
Catalog No. PRV9181 Supplier Promega Supplier No. V9181
Recombinant full-length human Aurora B was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. Aurora B is a member of the Aurora kinase family that associates with microtubules during chromosome movement and segregation.
  • Easily Screen and Profile Aurora B Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 Ea.
Content And Storage Components at different temperatures