Learn More
Promega ADP-Glo™ Kinase Assay + CLK1 Kinase Enzyme System
Catalog No. PRV4057 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRV4057 1 Ea.
1 options
Catalog No. PRV4057 Supplier Promega Supplier No. V4057
Add to Cart
Add to Cart
Recombinant human CLK1 (129-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CLK1, also known as STY, is a member of the CDC2-like (or LAMMER) family of dual specificity protein kinases.
  • Easily Screen and Profile CLK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 Ea.
Content And Storage Components at different temperatures
Kit Contents
  • CLK1 Kinase Enzyme System; (V4056) 1 x 10μg
  • ADP-Glo™ Kinase Assay; (V9101) 1 x 1,000 assays
Product Type Kinase Assay + Kinase Enzyme System