Description
- Easily Screen and Profile LCK Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Quantity | 1 Ea. |
| Content And Storage |
|
| Product Type | Kinase Assay + Kinase Enzyme System |