Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ADP-Glo™ Kinase Assay + PDGFRα (T674I) Kinase Enzyme System

Catalog No. PRV4487 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV4487 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV4487 Supplier Promega Supplier No. V4487
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human PDGFRα (Y674I) (550-end) point mutant was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

  • Easily Screen and Profile PDGFRα (T674I) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Quantity 1 Ea.
Content And Storage
  • PDGFRα (T674I) Kinase Enzyme System; (V4486) 1 x 10μg
  • ADP-Glo™ Kinase Assay; (V9101) 1 x 1,000 assays
Components at different temperatures
Product Type Kinase Assay + Kinase Enzyme System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.