Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ADP-Glo™ Kinase Assay with PI:3PS

Catalog No. PRV1782 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10,000 Assays
1000 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV1782 10,000 Assays
PRV1781 1000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV1782 Supplier Promega Supplier No. V1782
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The ADP-Glo™ Kinase assay and lipid kinase substrates PIP2:3PS and PI:3PS are components of the PI3K-Glo™ Class I Kinase Profiling System that are available for purchase separately.

  • Substrate and Assay Reagents for Detecting Class I PI3K Activity
  • Use in screening assays with purified PI3K Class I lipid kinases
  • Simple, scalable add-and-read assay format

Specifications

Quantity 10,000 Assays
Content And Storage
  • DP-Glo Reagent; (V912B) 1 x 50mL
  • Kinase Detection Buffer; (V913B) 1 x 100mL
  • Kinase Detection Substrate; (V914B) 1 x 1 each
  • Ultra Pure ATP, 10mM; (V915B) 1 x 5mL
  • ADP, 10mM; (V916B) 1 x 5mL
  • PI:3PS Substrate, 1mg/mL PI; (V171A) 10 x 0.5mL
  • Lipid Dilution Buffer, 10X; (V180A) 5 x 1mL
  • MgCl2, 1M; (V330A) 1 x 1mL
Upon receiving ADP-Glo™ Kinase Assay, remove ATP and store it below -65°C. Store the rest of the components at -30°C to -10°C. Before use, thaw all components completely at room temperature.
Format 384-well Plate
Product Type Lipid Kinase Assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.