Learn More
Promega ADP-Glo Kinase Assay with PIP2:3PS, 1,000 assays
Catalog No. PRV1791 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
Catalog No. Quantity
PRV1791 1000 Assays
1 options
Catalog No. PRV1791 Supplier Promega Supplier No. V1791
The ADP-Glo Kinase assay and lipid kinase substrates PIP2:3PS and PI:3PS are components of the PI3K-Glo Class I Kinase Profiling System that are available for purchase separately.
  • Substrate and Assay Reagents for Detecting Class I PI3K Activity
  • Use in screening assays with purified PI3K Class I lipid kinases
  • Simple, scalable “add−and−read” assay format
Specifications
Quantity 1000 Assays
Content And Storage Components at different temperatures