Learn More
Description
- Profile More Compounds In-House: ADP-Glo™ Kinase Assay + Kinase Enzyme System is optimized so that you are up and running in no time
- Complete Systems: Kinase Enzyme Systems include recombinant kinase enzyme, substrate appropriate for enzyme, reaction buffer, DTT and supplemental reagents as needed
- Obtain Reliable Results: Broad dynamic range, ease of use and better sensitivity obtained with ADP-Glo™ Kinase Assay result in less ambiguous data
Specifications
Specifications
| Quantity | 1 Ea. |
| Content And Storage | Store the kinase at -70°C and the rest of the components at -20°C. |
| Kit Contents |
|
| Product Type | Kinase Assay + Kinase Enzyme System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.