Learn More
Promega ADP-Glo™ Kinase Assay + TGFβR2 Kinase Enzyme System
Catalog No. PRV8301 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRV8301 1 Ea.
1 options
Catalog No. PRV8301 Supplier Promega Supplier No. V8301
Add to Cart
Add to Cart
Recombinant human TGF beta R2 (190-end) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. TGF beta R2 is a member of the TGF beta receptor subfamily and is a ser/thr protein kinase.
  • Easily Screen and Profile TGFβR2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 Ea.
Content And Storage Components at different temperatures
Kit Contents
  • TGFβR2 Kinase Enzyme System; (V3931) 1 x 10μg
  • ADP-Glo™ Kinase Assay; (V9101) 1 x 1,000 assays
Product Type Kinase Assay + Kinase Enzyme System