Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADP-ribosylarginine hydrolase Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP320168AF750
Description
ADP-ribosylarginine hydrolase Polyclonal antibody specifically detects ADP-ribosylarginine hydrolase in Human samples. It is validated for ELISASpecifications
ADP-ribosylarginine hydrolase | |
Polyclonal | |
ELISA | |
[Protein ADP-ribosylarginine] hydrolase, ADP-ribosylarginine hydrolaseARH1ADP-ribose-L-arginine cleaving enzyme, EC 3.2.2.19 | |
Produced in rabbits immunized with purified, recombinant human ADP-ribosylarginine hydrolase (Met1-Leu351; Accesion Number: NP_001116.1) | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
141 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction