Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADP-Sugar Pyrophosphatase/NUDT5 Antibody (10D2), Novus Biologicals™
SDP

Catalog No. NB298071 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298071 100 μL
NB298070 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298071 Supplier Novus Biologicals Supplier No. NBP326453100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ADP-Sugar Pyrophosphatase/NUDT5 Monoclonal antibody specifically detects ADP-Sugar Pyrophosphatase/NUDT5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen ADP-Sugar Pyrophosphatase/NUDT5
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 10D2
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 3.6.1.-, EC 3.6.1.13, hYSAH1, nucleoside diphosphate linked moiety X-type motif 5, Nucleoside diphosphate-linked moiety X motif 5, nudix (nucleoside diphosphate linked moiety X)-type motif 5, Nudix motif 5, YSA1, YSA1HADP-sugar pyrophosphatase
Host Species Rabbit
Immunogen A synthesized peptide derived from Human ADP-Sugar Pyrophosphatase/NUDT5 [UniProt Q9UKK9]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 11164
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.