Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADP-Sugar Pyrophosphatase/NUDT5 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300064APC
Description
ADP-Sugar Pyrophosphatase/NUDT5 Polyclonal antibody specifically detects ADP-Sugar Pyrophosphatase/NUDT5 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
ADP-Sugar Pyrophosphatase/NUDT5 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
EC 3.6.1.-, EC 3.6.1.13, hYSAH1, nucleoside diphosphate linked moiety X-type motif 5, Nucleoside diphosphate-linked moiety X motif 5, nudix (nucleoside diphosphate linked moiety X)-type motif 5, Nudix motif 5, YSA1, YSA1HADP-sugar pyrophosphatase | |
Produced in rabbits immunized with purified, recombinant Human ADP-Sugar Pyrophosphatase/NUDT5 (Uniprot#: Q9UKK9; Glu2-Phe219). | |
0.1 mL | |
Lipid and Metabolism | |
11164 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction