Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADP-Sugar Pyrophosphatase/NUDT5 Antibody (SR1511), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322427100UL
Description
ADP-Sugar Pyrophosphatase/NUDT5 Monoclonal antibody specifically detects ADP-Sugar Pyrophosphatase/NUDT5 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
ADP-Sugar Pyrophosphatase/NUDT5 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR1511 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
EC 3.6.1.-, EC 3.6.1.13, hYSAH1, nucleoside diphosphate linked moiety X-type motif 5, Nucleoside diphosphate-linked moiety X motif 5, nudix (nucleoside diphosphate linked moiety X)-type motif 5, Nudix motif 5, YSA1, YSA1HADP-sugar pyrophosphatase | |
A synthesized peptide derived from human ADP-Sugar Pyrophosphatase/NUDT5 (Uniprot #: Q9UKK9) | |
100 μg | |
Lipid and Metabolism | |
11164 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction