Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADP-Sugar Pyrophosphatase/NUDT5 Rabbit anti-Human, Mouse, Rat, Clone: 7Y9M2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | ADP-Sugar Pyrophosphatase/NUDT5 |
---|---|
Clone | 7Y9M2 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
ADP-Sugar Pyrophosphatase/NUDT5 Monoclonal antibody specifically detects ADP-Sugar Pyrophosphatase/NUDT5 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
ADP-Sugar Pyrophosphatase/NUDT5 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
11164 | |
IgG | |
Affinity purified |
7Y9M2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 3.6.1.-, EC 3.6.1.13, hYSAH1, nucleoside diphosphate linked moiety X-type motif 5, Nucleoside diphosphate-linked moiety X motif 5, nudix (nucleoside diphosphate linked moiety X)-type motif 5, Nudix motif 5, YSA1, YSA1HADP-sugar pyrophosphatase | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ADP-Sugar Pyrophosphatase/NUDT5 (Q9UKK9). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title