Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADPGK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560945
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560945 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560945 Supplier Invitrogen™ Supplier No. PA560945
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GPVGPKLHEL LDDNVFVPPE SLQEVDEFHL ILEYQAGEEW GQLKAPHANR FIFSHDLSNG AMNMLEVFVS SLEEFQPDLV VLSGLHMMEG QSKELQRKRL LEVVTSISDI PTGIPVHLEL ASMTNRELMS SIVH Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

The ADPGK gene encodes ADP-dependent glucokinase, an enzyme distinct from the ATP-dependent hexokinases as it employs adenosine diphosphate (ADP) rather than ATP to phosphorylate glucose to glucose-6-phosphate. This gene, initially identified in archaeal species, has been co-opted into mammalian systems where it plays roles primarily in glycolytic processes. Structurally, the mammalian ADPGK differs from its archaeal counterpart, exemplified by the rearrangement of its secondary structures, such as the alpha2 and alpha3 helices. These helices are replaced by an extended beta-strand region, indicating evolutionary adaptations for function in higher organisms like mice. Although its precise physiological role remains partially defined, ADPGK is notably expressed in both hematopoietic lineages and tumor tissues, suggesting potential significance in cancer biology and energy metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADPGK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADPGK
Gene Accession No. Q9BRR6
Gene Alias 2610017G09Rik; ADP dependent glucokinase; ADP-dependent glucokinase; ADPGK; Adp-gk; ATP-dependent glucokinase; C77577; EC 2.7.1.147; PSEC0260; RbBP-35; RGD1306103
Gene Symbols ADPGK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADPGK. Recombinant protein control fragment (Product #RP-91676).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83440
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.