Learn More
Invitrogen™ ADPGK Polyclonal Antibody

Description
Immunogen sequence: SPGNGKDHSIL HSRNDLEEAF IHFMGKGAAA ERFFSDKETF HDIAQVASEF PGAQHYVGGN AALIGQKFAA NSDLKVLLC Highest antigen sequence identity to the following orthologs - mouse 90%, rat 89%.
Specifications
Specifications
| Antigen | ADPGK |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ADPGK |
| Gene Accession No. | Q9BRR6 |
| Gene Alias | 2610017G09Rik; ADP dependent glucokinase; ADP-dependent glucokinase; ADPGK; Adp-gk; ATP-dependent glucokinase; C77577; EC 2.7.1.147; PSEC0260; RbBP-35; RGD1306103 |
| Gene Symbols | ADPGK |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.