Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADPRHL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555521 Supplier Thermo Scientific Supplier No. PA555521
Add to Cart
Add to Cart

ADPRHL2 Polyclonal Antibody

ADPRHL2 Polyclonal Antibody for Western Blot, ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ADPRHL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADPRHL2
Gene Accession No. Q9NX46
Gene Alias [Protein ADP-ribosylarginine] hydrolase-like protein 2; [Protein ADP-ribosylserine] hydrolase; ADPRHL2; ADP-ribose glycohydrolase ARH3; ADP-ribose glycohydrolase ARH3; poly(ADP-ribose) glycohydrolase ARH3; ADP-ribosylarginine hydrolase like 2; ADP-ribosylhydrolase 2; ADP-ribosylhydrolase 3; ADP-ribosylhydrolase like 2; ADP-ribosylserine hydrolase; ADPRS; AI836109; ARH3; ecto ADP-ribosylhydrolase; LOW QUALITY PROTEIN: poly(ADP-ribose) glycohydrolase ARH3; O-acetyl-ADP-ribose deacetylase ARH3; Poly(ADP-ribose) glycohydrolase ARH3; protein ADP-ribosylarginine hydrolase-like protein 2
Gene Symbols ADPRHL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADPRHL2. Recombinant protein control fragment (Product #RP-94093).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54936
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.